Thermo Scientific™ Nunc™ EasYDish™ Dishes
Next generation cell culture dishes that improve handling, stacking and transporting of cell cultures in your lab.
Overview
86.64 GBP valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Additional Details:
Additional Details: Weight: 3.00000kg
Product Code. 15873937
Quantity Price
1 £ 200.0 / Pack of 280
In stock, ready to ship: 3
In stock, ready to ship: 489
Add to basket
  • Description and Specification
  • Alternative Products
  • Special Offers
Description and Specification
Specification
Closure Included Closure Included
Closure Material Polystyrene
Description Nunc EasYDish
Bottom Shape Flat
Capacity (Metric) 5 mL (Well)
Culture Area 21.5 sq. cm
Gridded Not Gridded
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Nunclon Delta
Type Cell Culture Dish
Volume (Metric) Working 5 mL
Includes Lid
No. per Case 280
No. per Pack 10
Height (Metric) 16 mm
Diameter (Metric) 60 mm
Beveled Grip Ring
  • Ensures lid stays in position when dish is being handled
  • Makes it easier to grasp and manage with a gloved hand
Proprietary Notch Design
  • Maximizes ventilation while maintaining sterility
  • Restricts the movement of the lid ensuring secure lid-to-dish fitting
  • Increases the physical strength of the dish
Raised Outer Edge on Dish Lid
  • Allows stabled stacking during transportation of cell cultures
Orientation Marks on Dish Bottom
  • Simplifies locating of cells under the microscope
Writing Area
  • Improves traceability of cell cultures
Re-Sealable Packaging
  • Promotes cleanliness of opened packs
Specifications:
  • Material: Polystyrene, USP VI
  • Contaminant Free: RNase/DNase free, Human DNA free, non-cytotoxic, non-pyrogenic
  • Sterility: Gamma Irradiation to SAL of 10-6
  • Shelf Life: 5 years from manufacturing date
  • Certificate of Quality in every case
  • Surface Treatment: Nunclon Delta for adherent cell cultures