Eppendorf™ Centrifuge 5430/5430 R
The unique, cross-over Eppendorf Centrifuge 5430 and 5430 R combine the small footprint of a microcentrifuge with the versatility of a multipurpose centrifuge. Extremely compact, they can accommodate rotors for microplates and 15/50 mL conical tubes as well as Eppendorf tubes and PCR strips.
6804.00 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Use promo code "25339" to get your promotional price.
Additional Details:
Weight:
57.00000kg
Specification
Capacity | 30 x 2 mL |
---|---|
Max. Speed | 17,500 rpm |
Max. RCF | 30,130 x g |
Refrigerated | Yes |
Display | Large, brightly lit LCD |
Height (English) Exterior | 11.7 in. |
Height (Metric) Exterior | 29 cm |
Noise Level | <54 dB(A) with Rotor F-45-30-11 |
Safety Features | Lid Lock, steel armoured safety chamber, view port for speed validation, lid emergency release, automatic rotor recognition, certified aerosol-tight rotor lids, automatic imbalance recognition |
Timer | 30 sec. to 99:59 hrs., with continuous run function |
Width (English) Exterior | 15 in. |
Width (Metric) Exterior | 38 cm |
Wattage | 1.050 W |
Certifications/Compliance | UL (120 V), CAN/CSA (120 V), CE, RoHS, IVD for certain catalog numbers |
Includes | With Rotor FA-45-30-11 incl. rotor lid |
Dimensions (L x W x H) Exterior | 380 x 640 x 290 mm |
Electrical Requirements | 230 V, 50/60 Hz (GB) |
Controller Type | Rotary Knob |
Rotor Included | Yes |
Standards | EN ISO 12100, EN 50581, EN 61010-1, IEC 61010-2-020, EN 61326-1, EN 55011, EN 378-1, EN 378-2, 2 EN 50581 (refer to manual for more details) |
Temperature Range | -11°C to +40°C |
Frequency | 50/60 Hz |
Voltage | 230 V |
Model | Centrifuge 5430 R |