Use promo code "24140" to get your promotional price.
Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
Use promo code "24140" to get your promotional price.
| Quantity | 4 L |
|---|
Chemical Identifiers
| CAS | 64-17-5 |
|---|---|
| Molecular Formula | C2H6O |
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| PubChem CID | 702 |
| ChEBI | CHEBI:16236 |
| IUPAC Name | ethanol |
| SMILES | CCO |
Specification
| Chemical Name or Material | Ethanol |
|---|---|
| Melting Point | -114°C |
| Name Note | Absolute (200 Proof) |
| CAS | 64-17-5 |
| Color | Colorless |
| Quantity | 4 L |
| Flash Point | 12°C (53.6°F) |
| Identification | Pass Test |
| Infrared Spectrum | Conforms to reference |
| Physical Form | Liquid |
| Purity Grade Notes | DNase-, RNase- and Protease-Free |
| Residue after Evaporation | 0.001% max. |
| Assay Percent Range | Assay: ≥99.5% (v/v) |
| Titratable Acid | 0.0005 mEq /g max. |
| Molecular Formula | C2H6O |
| Titratable Base | 0.0002 mEq /g max. |
| Linear Formula | C2H5OH |
| Water | 0.2% max. |
| Merck Index | 15, 3814 |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Synonym | ethyl alcohol,alcohol,methylcarbinol,grain alcohol,ethyl hydroxide,ethyl hydrate,algrain,alcohol,anhydrol,tecsol |
| Boiling Point | 78°C |
| InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
| SMILES | CCO |
| IUPAC Name | ethanol |
| Molecular Weight (g/mol) | 46.069 |
| PubChem CID | 702 |
| ChEBI | CHEBI:16236 |
| Formula Weight | 46.07 |
| Percent Purity | ≥99.5% |
| Grade | Molecular Biology |
[Molecular Biology Grade Ethanol]
[Danger]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H301, sequence=2, description=Toxic if swallowed.}, {code=H311, sequence=3, description=Toxic in contact with skin.}, {code=H331, sequence=4, description=Toxic if inhaled.}, {code=H370, sequence=5, description=Causes damage to organs.}]
[{code=P301+P310, sequence=51, description=IF SWALLOWED: Immediately call a POISON CENTER/doctor /}, {code=P303+P361+P353, sequence=52, description=IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.}, {code=P321, sequence=53, description=Specific treatment (see on this label).}, {code=P330, sequence=54, description=Rinse mouth.}, {code=P362+P364, sequence=55, description=Take off contaminated clothing and wash it before reuse.}, {code=P405, sequence=56, description=Store locked up.}, {code=P501b, sequence=57, description=Dispose of contents/container in accordance with local/regional/national/international regulations.}]
[{code=MIXTURE LIST, sequence=1, description=Contain: ethanol}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS06, sequence=2, description=SKULL & CROSSBONES}, {code=GHS08, sequence=3, description=HEALTH HAZARD}]
Product Identifier
- Molecular Biology Grade Ethanol
Signal Word
- Danger
Hazard Statement
- H225- Highly flammable liquid and vapour.
- H301- Toxic if swallowed.
- H311- Toxic in contact with skin.
- H331- Toxic if inhaled.
- H370- Causes damage to organs.
Precautionary Statement
- P301+P310- IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P321- Specific treatment (see on this label).
- P330- Rinse mouth.
- P362+P364- Take off contaminated clothing and wash it before reuse.
- P405- Store locked up.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental Information
- MIXTURE LIST- Contain: ethanol