Ecolab Filtered Klercide 70/30 IPA Blended with Deionized Water
This Klercide 70/30 IPA blended with deionized water is cleanroom filtered for use in lower grade areas.
Additional Details:
Additional Details: CAS Number: 67-63-0 Weight: 5.26500kg Transport: UN number : 1219 Chem class : 3 Pack group : II
Product Code. 17815497
Quantity Price
1 £ 245.44 / 6 L
In stock, ready to ship: 18
Add to basket
  • Description and Specification
  • Alternative Products
  • Safety and Handling
Description and Specification
Specification
Product Type Disinfectant
Form Liquid
pH Neutral
Sterility Sterile
Volume (Metric) 6 x 1 L
  • BPR approved
  • Economical disinfection suitable for use in lower grade areas
  • 0.2 micron filtered, then filled and double bagged in a Grade C (ISO Class 7) cleanroom
  • Supplied in spray bottle which incorporates the patented SteriShield Delivery System and an adjustable trigger spray. The unique way in which the SteriShield Delivery System works, as a closed system, ensures that the integrity of the contents is preserved throughout use
  • Certificates of Analysis are available for every batch
  • Full supporting technical documentation is available
Alternative Products
Safety and Handling
[Klercide 70/30 IPA]
[Danger]
[{code=EYE DAM. 2, sequence=1, description=Serious eye damage/eye irritation Category 2}, {code=FLAM. LIQ. 2, sequence=2, description=Flammable liquid Category 2}, {code=STOT SE 3, sequence=3, description=Specific target organ toxicity Category 3}]
[{code=H225, sequence=1, description=Highly flammable liquid and vapour.}, {code=H319, sequence=2, description=Causes serious eye irritation.}, {code=H336, sequence=3, description=May cause drowsiness or dizziness.}, {code=EUH066, sequence=91, description=Repeated exposure may cause skin dryness or cracking.}]
[{code=P210, sequence=51, description=Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.}, {code=P280, sequence=52, description=Wear protective gloves/protective clothing/eye protection/face protection.}]
[{code=MIXTURE LIST, sequence=1, description=Contains : Isopropanol}]
[{code=GHS02, sequence=1, description=FLAME}, {code=GHS07, sequence=2, description=EXCLAMATION MARK}]
Product Identifier
  • Klercide 70/30 IPA
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225- Highly flammable liquid and vapour.
  • H319- Causes serious eye irritation.
  • H336- May cause drowsiness or dizziness.
  • EUH066- Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280- Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental Information
  • MIXTURE LIST- Contains : Isopropanol