Honeywell Research Chemicals 2,2,4-Trimethylpentane, ≥99%, Reagent Grade, Honeywell™

Reagent Grade,≥99%

Overview
Code: NEW

Additional Details:
Additional Details: CAS Number: 540-84-1 Weight: 0.34600kg Transport: UN number : 1262 Chem class : 3 Pack group : II


Quantity 500 mL
Packaging Glass Bottle

Product Code. 15602800

 N/A
N/A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


  • Description and Specification
  • Alternative Products
  • Documentation
Description and Specification

Chemical Identifiers

CAS 540-84-1
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00008943
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
PubChem CID 10907
ChEBI CHEBI:62805
IUPAC Name 2,2,4-trimethylpentane
SMILES CC(C)CC(C)(C)C

Specification

Chemical Name or Material 2,2,4-Trimethylpentane
Melting Point -107°C
CAS 540-84-1
Color Colorless
Density 0.692g/mL at 25°C
Boiling Point 98°C to 99°C
Quantity 500 mL
Explosion Limit 0.06
Flash Point −12°C (10.4°F)
Ignition Point 745°F
Packaging Glass Bottle
Physical Form Liquid
Molecular Formula C8H18
Linear Formula (CH3)2CHCH2C(CH3)3
MDL Number MFCD00008943
Vapor Pressure 41mmHg (21°C)
UN Number UN1262
Beilstein 1696876
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
Synonym isooctane,isobutyltrimethylmethane,pentane, 2,2,4-trimethyl,iso-octane,2,4,4-trimethylpentane,unii-qab8f5669o,ch3 2chch2c ch3 3,2, 2, 4-trimethylpentane,i-octane,isobutyltrimethylethane
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.232
PubChem CID 10907
ChEBI CHEBI:62805
Formula Weight 114.23g/mol
Percent Purity ≥99%
Grade ReagentPlus™

  • ≥95% for general laboratory use.

Alternative Products
Documentation

Technical documents