Learn More
A2BP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22647-1-AP-20UL
 View more versions of this product
Product Code. 16832025
  • 145.00 € / 20µL
Explore more special offers
Description
Description
This is a rabbit polyclonal antibody raised against the N terminus of human A2BP1.
Ataxin-2 binding protein 1 has an RNP motif that is highly conserved among RNA-binding proteins. This protein binds to the C-terminus of ataxin-2 and may contribute to the restricted pathology of spinocerebellar ataxia type 2 (SCA2). Ataxin-2 is the gene product of the SCA2 gene which causes familial neurodegenerative diseases. Ataxin-2 binding protein 1 and ataxin-2 are both localized in the trans-Golgi network. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been found but their full length nature has not been determined.
Specifications
Specifications
A2BP1
Polyclonal
Unconjugated
RBFOX1
A2BP, A2BP1, ataxin 2 binding protein 1, Fox 1 homolog A, FOX1, HRNBP1, RBFOX1
Rabbit
Antigen Affinity Chromatography
RUO
268859, 54715
-20°C
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9JJ43, Q9NWB1
RBFOX1
A2BP1 Fusion Protein Ag18527
20 μL
Primary
Human, Mouse