Proteintech ABCD1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Additional Details:
Additional Details: Weight: 0.00010kg



Product Code. 16858043

Quantity Price
1 410.0  € / 150µL
In stock, ready to ship: 4
Add to basket


Description and Specification

Specification

Antigen ABCD1
Applications Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ABCD1
Gene Accession No. P33897, P48410
Gene Alias ABC42, ABCD1, Adrenoleukodystrophy protein, ALD, ALDP, AMN
Gene Symbols Abcd1
Host Species Rabbit
Immunogen ABCD1 Fusion Protein Ag1635
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11666, 215, 363516
Target Species Human,Mouse,Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG

This gene is a member of the UBA domain family, whose members include proteins having connections to ubiquitin and the ubiquitination pathway. The ubiquitin associated domain is thought to be a non-covalent ubiquitin binding domain consisting of a compact three helix bundle. This particular protein originates from a gene locus in a refined region on chromosome 9 undergoing loss of heterozygosity in nasopharyngeal carcinoma (NPC). Taking into account its cytogenetic location, this UBA domain family member is being studies as a putative target for mutation in nasopharyngeal carcinomas. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene.