Learn More
Miele Professional™ Accessory Booster Pump
Rec in/out water inlet and drain for PLW 8636
713.00 € - 1581.00 €
Specifications
Product Type Booster Pump
For Use With (Equipment) Washer-Disinfectors
Quantity 1 Ea.
View More Specs
Products 2
Product Code Brand Rack Type Price Quantity & Availability  
Product Code Brand Rack Type Price Quantity & Availability  
30533240
View Documents
Miele Professional™
12426490
Rec in
713.00 €
1 sheet
View alternative products
Save Up to 
»
30533236
View Documents
Miele Professional™
12426550
Rec out
1581.00 €
1 sheet
View alternative products
Save Up to 
»
Description
Description
  • Rec in/out water inlet and drain for PLW 8636
  • Required in order to pump a medium out of a buffer container
  • For use with an existing on-site recycling tank
  • Can be used with existing on-site container, e.g. IBC tank
  • Installed by Customer Service
Specifications
Specifications
Booster Pump
Washer-Disinfectors
1 Ea.