Product Code | Brand | Quantity | Packaging | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
---|---|---|---|---|---|---|---|---|---|
10666821
|
JT Baker
6052.1000
|
1 L | Glass bottle |
72.35 €
1 L |
|
||||
15578364
|
JT Baker
6052.1000PE
|
1 L | Plastic bottle |
67.1 €
1 L |
|
||||
10371411
|
JT Baker
6052.2500
|
2.5 L | Glass bottle |
152.0 €
2.50 L |
|
Acetic acid glacial 99-100%, BAKER ANALYZED™ ACS, J.T. Baker™
Exceeds ACS Specifications
67.1 € - 152.0 €
Chemical Identifiers
CAS | 64-19-7 |
---|---|
Molecular Formula | C2H4O2 |
Molecular Weight (g/mol) | 60.05 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
IUPAC Name | acetic acid |
SMILES | CC(O)=O |
Specification
CAS | 64-19-7 |
---|---|
Color | Max. 10 (APHA) |
Assay Percent Range | min. 99.7% |
Molecular Formula | C2H4O2 |
MDL Number | MFCD00036152 |
InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
SMILES | CC(O)=O |
IUPAC Name | acetic acid |
Molecular Weight (g/mol) | 60.05 |
Quantity | 1 L |
Packaging | Glass bottle |
Residue after Evaporation | max. 0.001% |
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226- Flammable liquid and vapour.
- H314- Causes severe skin burns and eye damage.
Precautionary Statement
- P210- Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353- IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338- IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310- Immediately call a POISON CENTER/doctor/.
- P405- Store locked up.
- P501b- Dispose of contents/container in accordance with local/regional/national/international regulations.