Product Code | Brand | Quantity | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
---|---|---|---|---|---|---|---|---|
16890135
|
Proteintech
20578-1-AP-20UL
|
20 μL |
145.0 €
20µL |
|
||||
16880135
|
Proteintech
20578-1-AP-150UL
|
150 μL |
410.0 €
150µL |
|
APOB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.0 € - 410.0 €
Specification
Antigen | APOB |
---|---|
Concentration | 0.18 mg/mL |
Applications | Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Specification
Antigen | APOB |
---|---|
Concentration | 0.18 mg/mL |
Applications | Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Host Species | Rabbit |
Regulatory Status | RUO |
Target Species | Mouse,Human |
Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
Gene Accession No. | E9Q414, P04114 |
Gene Alias | Apo B 100, Apo B 48, APOB, ApoB100, Apolipoprotein B, Apolipoprotein B 100, FLDB |
Gene ID (Entrez) | 238055, 338 |
Gene Symbols | APOB |
Immunogen | Peptide |
Isotype | IgG |
Primary or Secondary | Primary |
Purification Method | Antigen Affinity Chromatography |
Content And Storage | -20°C |
Product Type | Antibody |
Gene | APOB |