Learn More
ATG2A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 23226-1-AP-20UL
 View more versions of this product
Product Code. 16862245
  • 145.00 € / 20µL
Description
Required for both autophagosome formation and regulation of lipid droplet morphology and dispersion.
Specifications
Specifications
ATG2A
Polyclonal
Unconjugated
ATG2A
ATG2A, KIAA0404
Rabbit
Antigen Affinity Chromatography
RUO
23130
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q2TAZ0
ATG2A
ATG2A Fusion Protein Ag19709
20 μL