Proteintech beta Tubulin Mouse anti-C. elegans, Human, Mouse, Porcine, Rat, Zebrafish, Clone: 1D4A4, Proteintech
Mouse Monoclonal Antibody
Additional Details:
Weight:
0.00010kg
Specification
Antigen | beta Tubulin |
---|---|
Applications | Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot |
Applications | Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot |
Applications | Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot |
Applications | Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot |
Applications | Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot |
Classification | Monoclonal |
Clone | 1D4A4 |
Concentration | 1.43 mg/mL |
Conjugate | Unconjugated |
Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
Gene | TUBB3 |
Gene Accession No. | Q13509, Q4QRB4, Q9ERD7 |
Gene Alias | beta tubulin, MC1R, TUBB, Tubulin-beta |
Gene Symbols | TUBB3 |
Host Species | Mouse |
Immunogen | beta Tubulin Fusion Protein Ag0117 |
Purification Method | Protein A |
Quantity | 150 μL |
Regulatory Status | RUO |
Primary or Secondary | Primary |
Gene ID (Entrez) | 102154194, 10381, 106509340, 22152, 246118 |
Target Species | C. elegans,Human,Mouse,Pig,Rat,Zebrafish |
Target Species | C. elegans,Human,Mouse,Pig,Rat,Zebrafish |
Target Species | C. elegans,Human,Mouse,Pig,Rat,Zebrafish |
Target Species | C. elegans,Human,Mouse,Pig,Rat,Zebrafish |
Target Species | C. elegans,Human,Mouse,Pig,Rat,Zebrafish |
Target Species | C. elegans,Human,Mouse,Pig,Rat,Zebrafish |
Content And Storage | -20°C |
Product Type | Antibody |
Form | Liquid |
Isotype | IgG2a |