missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium carbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance), Honeywell Fluka™
Shop All Honeywell Fluka Products

Click to view available options
Quantity:
1 kg
2.5 kg
25 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Quantity | 6 x 1 kg |
Linear Formula | CaCO3 |
UN Number | NONH for all modes of transport |
IUPAC Name | calcium carbonate |
Formula Weight | 100.09 |
Percent Purity | 98.5 to 100.5% (based on anhydrous substance) |
Grade | puriss. |
Density | 2.93g/mL at 25°C |
Chemical Name or Material | Calcium Carbonate(Caco3), Meets analytical specification of Ph. Eur.BP, USP, FCC |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction