Proteintech Cd31 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Additional Details:
Additional Details: Weight: 0.00010kg



Product Code. 16835985

Quantity Price
1 410.0  € / 150µL
In stock, ready to ship: 8
Add to basket


Description and Specification

Specification

Antigen Cd31
Applications Western Blot,Immunohistochemistry,Immunofluorescence
Classification Polyclonal
Concentration 267 μg/mL
Conjugate Unconjugated
Host Species Rabbit
Quantity 150 μL
Primary or Secondary Primary
Gene ID (Entrez) 18613
Target Species Mouse,Human
Isotype IgG

The Cd31 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Cd31. This antibody recognizes mouse, human antigen. The Cd31 antibody has been validated for the following applications: IHC, WB, ELISA analysis.