Learn More
Cd31 Rabbit anti-Mouse, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28083-1-AP-20UL
 View more versions of this product
Product Code. 16845985
  • 145.00 € / 20µL
Description
The Cd31 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of mouse Cd31. This antibody recognizes mouse, human antigen. The Cd31 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
Cd31
Polyclonal
Unconjugated
20 μL
18613
IgG
Western Blot, Immunohistochemistry, Immunofluorescence
267 μg/mL
Rabbit
Primary
Mouse, Human