missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate
Color: Clear; Sterility: Sterile; Lid: Without Lid
Specifications
Format | Standard |
---|---|
Lid | Without Lid |
Culture Area | 34 mm2 |
Array | 8 x 12 |
Color | Clear |
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Compatible with automated systems
- Alphanumeric well coding
- High clarity of microplates for optimal microscopic examinations
- Stackable
- Individu
Specifications
Standard | |
34 mm2 | |
Clear | |
96 | |
Cell Culture | |
4800 x g (swinging bucket rotor) |
Without Lid | |
8 x 12 | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) | |
Chimney |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title