missing translation for 'onlineSavingsMsg'
Learn More

Citric Acid, 99.5 to 100.5% (based on Anhydrous substance), Honeywell Fluka™
BIOPHARMA

Product Code. p-7071950 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
25 kg
6 x 1 kg
6 x 2.5 kg
Unit Size:
100g
1kg
2.5kg
25kg
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15662880 100 g 100g
15672880 1 kg 1kg
15682880 2.5 kg 2.5kg
15602890 6 x 1 kg Pack of 6
15612890 6 x 2.5 kg Pack of 6
15692880 25 kg 25kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15662880 Supplier Honeywell-Fluka Supplier No. 27109100G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Meets analytical specification of BP, Ph. Eur., USP, E330, Anhydrous, 99.5-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-92-9
Molecular Formula C6H8O7
Molecular Weight (g/mol) 192.12
MDL Number MFCD00011669
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
PubChem CID 311
ChEBI CHEBI:30769
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O

Specifications

Linear Formula HOC(COOH)(CH2COOH)2
UN Number NONH for all modes of transport
Beilstein 782061
Solubility Information Soluble in water, 383g/L at 25°C, Sparingly soluble in Diethyl Ether, Freely soluble in alcohol
Formula Weight 192.12g/mol
Percent Purity 99.5 to 100.5% (based on anhydrous substance)
Quantity 100 g
Explosion Limit 8% (65°F)
Physical Form Anhydrous
Residue after Ignition ≤0.05% (as SO4)
Chemical Name or Material Citric Acid, Meets analytical specification of Ph. Eur.BP, USP
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Citric acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.