Proteintech COMT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Additional Details:
Weight:
0.00010kg
Specification
Antigen | COMT |
---|---|
Applications | Immunoprecipitation,Immunofluorescence,Immunocytochemistry,Immunohistochemistry (Paraffin),Flow Cytometry |
Classification | Polyclonal |
Concentration | 0.19 mg/mL |
Conjugate | Unconjugated |
Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
Gene | COMT |
Gene Accession No. | O88587, P21964, P22734 |
Gene Alias | catechol O methyltransferase, COMT |
Gene Symbols | COMT |
Host Species | Rabbit |
Immunogen | COMT Fusion Protein Ag6502 |
Purification Method | Antigen Affinity Chromatography |
Quantity | 20 μL |
Regulatory Status | RUO |
Primary or Secondary | Primary |
Gene ID (Entrez) | 12846, 1312, 24267 |
Target Species | Human,Mouse,Rat |
Content And Storage | -20°C |
Product Type | Antibody |
Form | Liquid |
Isotype | IgG |