missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper Standard For ICP, 10,000μg/mL in 5% HNO3, Assurance Grade SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
98.85 € - 219.00 €
Chemical Identifiers
CAS | 7440-50-8, 7697-37-2, 7732-18-5 |
---|---|
Molecular Formula | Cu |
Molecular Weight (g/mol) | 63.55 |
MDL Number | MFCD00010965 |
InChI Key | RYGMFSIKBFXOCR-UHFFFAOYSA-N |
PubChem CID | 23978 |
ChEBI | CHEBI:30052 |
IUPAC Name | copper |
SMILES | [Cu] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10361504
|
SPEX Certiprep
PLCU2-3Y |
125 mL | Triple-leached LDPE bottle |
98.85 €
125mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10111912
|
SPEX Certiprep
PLCU2-3X |
500 mL | Triple-leached LDPE bottle |
219.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
Cu | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7440-50-8, 7697-37-2, 7732-18-5 | |
63.55 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
CHEBI:30052 | |
[Cu] |
Cu | |
MFCD00010965 | |
23978 | |
copper |
Specifications
7440-50-8, 7697-37-2, 7732-18-5 | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
MFCD00010965 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
63.55 | |
CHEBI:30052 | |
5% HNO3 | |
Copper Standard |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
Cu | |
AA, ICP | |
[Cu] | |
copper | |
23978 | |
Certified Reference Material | |
Assurance |
Safety and Handling
Causes skin irritation, serious eye irritation
missing translation for 'dotInformation' : Hazardous
Recommended Storage : Ambient
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : nitric acid, copper
Product intended for laboratory use only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title