Learn More
Corning™ Black Phenolic Threaded Screw Caps
Resist effects of temperature and steam in autoclaving
Brand: Corning™ 9999-40
This item is not returnable.
View return policy
Description
These caps are fabricated of a resin specially formulated to resist the effects of temperature and steam in autoclaving. Caps have a glued-in, white rubber liner.
Specifications
Black | |
72/Pk. | |
40-400 | |
12.5 mm |
Phenolic Resin | |
Rubber | |
Autoclavable | |
Screw Cap |