Honeywell KCL™ Dermatril™ P 743 Gloves

Honeywell KCL™ Dermatril™ P 743 Gloves feature chemical and mechanical resistance and are approved for food processing.

Overview
Code: NEW

Additional Details:
Additional Details: Weight: 0.60000kg



Product Code. 15350392

Quantity Price
1 19.85  € / Pack of 50
In stock, ready to ship: 39
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Specification

Certifications/Compliance ASTM 1671:2007
Certifications/Compliance (Europe) Approved for food processing according to EU 1935/2004. EC Category PSA 3
Color Blue
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size 10
Thickness (Metric) 0.2 mm
Quantity 50/Pk
Resistant To Variety of cytotoxic agents, virus-resistant to ASTM F1671: 2007
Length (Metric) 270 to 290 mm

  • Disposable protective glove according to EN 374
  • The chemical as well as the mechanical resistance are increased compared to the Dermatril™ 740
  • Can be used longer and offers protection against a wider range of hazardous substances.
  • Approved for food processing according to EU 1935/2004
  • Food grade through RAL approval
  • Silicone free glove surface
  • Meets the requirements of EN 455
  • Medical device according to directive 93/42/EWG
  • Free from natural latex
  • Resistant against a multitude of cytostatic drugs virus-resistant according to ASTM 1671:2007
  • High tactility
  • Excellent wearing comfort
  • Increased tear resistance
  • Improved grip
  • Good chemical resistance
  • Snug fit

Alternative Products