Epredia™ Disposable Base Molds
Use Epredia™ Disposable Base Molds with all standard-size cassettes.
Overview
Code: A0
Additional Details:
Additional Details: Weight: 0.01000kg
Product Code. 12580107
 N/A
N/A
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description and Specification
Specification
Product Type Base Mold
Material Plastic
Dimensions (L x W) 7 x 7 mm
Disposable Single-use
  • Cleaning associated with metal molds is not required
  • Transparency facilitates tissue orientation
Ordering Information:
  • Sold in cases of four boxes.