Learn More
ER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21244-1-AP-20UL
 View more versions of this product
Product Code. 16830865
  • 145.00 € / 20µL
Description
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.
Specifications
Specifications
ER
Polyclonal
Unconjugated
ESR1
ER, ER alpha, Era, ESR, ESR1, ESRA, Estradiol receptor, Estrogen receptor, estrogen receptor 1, NR3A1
Rabbit
Antigen Affinity Chromatography
RUO
13982, 2099, 24890
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.5 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P03372, P06211, P19785
ESR1
ER Fusion Protein Ag15738
20 μL
Primary
Mouse, Rat, Human