Learn More
EXOG Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 21523-1-AP-150UL
 View more versions of this product
Product Code. 16821115
  • 410.00 € / 150µL
Description
This gene encodes an endo/exonuclease with 5′-3′ exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5′ end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18.
Specifications
Specifications
EXOG
Polyclonal
Unconjugated
EXOG
Endo G like 1, ENDOGL1, ENDOGL2, Endonuclease G like 1, ENGL, ENGL a, ENGL B, EXOG, Nuclease EXOG, mitochondrial
Rabbit
Antigen Affinity Chromatography
RUO
208194, 301062, 9941
-20°C
Liquid
Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8C163, Q9Y2C4
EXOG
EXOG Fusion Protein Ag16122
150 μL
Primary
Human, Mouse, Rat