Learn More
FABP5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12348-1-AP-20UL
 View more versions of this product
Product Code. 16870014
  • 145.00 € / 20µL
Description
Fatty acid-binding proteins, designated FABPs, are a family of homologous cytoplasmic proteins that are expressed in a highly tissue-specific manner and play an integral role in the balance between lipid and carbohydrate metabolism. FABPs mediate fatty acid (FA) and/or hydrophobic ligand uptake, transport and targeting within their respective tissues. The mechanisms underlying these actions can give rise to both passive diffusional uptake and protein-mediated transmembrane transport of FAs. FABPs are expressed in adipocytes (A-FABP), brain (B-FABP), epidermis (E-FABP, also designated psoriasis-associated FABP or PA-FABP), muscle and heart (H-FABP, also designated mammary-derived growth inhibitor or MDGI), intestine (I-FABP), liver (L-FABP), myelin (M-FABP) and testis (T-FABP). Epithelial fatty acid-binding protein (E-FABP) binds stearic acid and may play a role in keratinocyte differentiation. E-FABP is upregulated in rat dorsal root ganglia after sciatic nerve crush and in differentiating neurons during development.
Specifications
Specifications
FABP5
Polyclonal
Unconjugated
FABP5
E FABP, EFABP, FABP5, Fatty acid binding protein 5, PA FABP, PAFABP
Rabbit
Antigen Affinity Chromatography
RUO
140868, 16592, 2171
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry
0.43 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P55053, Q01469, Q05816
FABP5
FABP5 Fusion Protein Ag3005
20 μL
Primary
Human, Mouse, Rat