missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Maxi Pipet Tips
GREENER_CHOICE

Product Code. 17838983 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
152 mm
161 mm
Unit Size:
Pack of 100
Pack of 200
Pack of 250
This item is not returnable. View return policy

Product Code. 17838983

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with large volume pipets

  • Designed for use when transferring large sample volume
  • Advanced molding technology ensure tips are straight
  • Alternative option to serological pipets
  • Nonsterile; certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 10 mL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Length (Metric) 152 mm
Format Bulk
Tip Style Macro
For Use With (Equipment) Fisherbrand II, Thermo Scientific
Product Line Maxi
Material Polypropylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.