Fisherbrand™ Polypropylene Disposable Beakers
Highly resistant to strong acids, organic solvents and other common laboratory reagents
Additional Details:
Weight:
1.20000kg
Specification
| Capacity (Metric) | 250 mL |
|---|---|
| Material | Polypropylene |
| For Use With (Application) | Highly Resistant to Strong Acids, Organic Solvents and Other Common Laboratory Reagents |
| Includes | Spout |
| Increments | 10 mL |
| Type | Beaker |
| Autoclavable | Autoclavable |
| Disposable | Single-use |
| Graduated | Yes |