missing translation for 'onlineSavingsMsg'
Learn More

Folin-Ciocalteu's Reagent, Reagecon
BIOPHARMA

Product Code. 16440704 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 16440704

Brand: Reagecon FOCIRE01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Description Liquid reagent comes in amber glass Winchester bottle
Product Type Folin-Ciocalteu's reagent
Quantity 100 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.