Buchi Column, Prep; Buchi; Used in combination with the injection unit for loading dry samples or barely soluble samples; Volumes up to 53mL
Overview
Column, Prep; Buchi; Used in combination with the injection unit for loading dry samples or barely soluble samples; Volumes up to 53mL
Code: A0
Additional Details:
Additional Details: Weight: 1.33000kg
Product Code. 11987208
 N/A
N/A