Learn More
3M™ Goggle Gear™ Safety Goggles, Anti-Fog / Anti-Scratch Coating (K&N)
Click to view available options
Lens Tint:
Gray
Transparent
Unit Size:
Pack of 10
Description
- Modern slim-line and lightweight design for an excellent fit
- Adjustable Neoprene headband for easy cleaning and sterilisation
- Pivoting ratchet for convenient personal adjustment
- 3M™ Scotchgard™ Anti-Fog Coating provides superior anti-fog and anti-scratch properties
- Coating withstands washing, soaking in bleach or using alcohol wipes without losing performance
- Special 3M™ Goggle Gear prescription insert available
- Offers excellent protection against UV radiation
- Lens Marking: 2C-1.2 3M 1 BT KN
- Frame Marking: 3M EN 166 3 4 BT CE
- Protection against large dust particles (3)
- Protection against liquid droplets (4)
Specifications
Specifications
Lens Coating | Anti-fog |
Lens Tint | Transparent |
Frame Color | Gray and Red |
Model | GG501NSGAF-EU |
Quantity | 10/Cs. |
Lens Material | Polycarbonate |
Frame Type | Goggle |