missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Graduated SteriWare Beakers

This range of graduated beakers are crucial in the production of drugs, chemicals and cosmetics. Individually bagged to prevent cross contamination, these beakers also have a pouring spout to ensure the convenient handling of liquids.

Brand:  Sampling Systems™ 8008P-50

 View more versions of this product

Product Code. 17922062

  • ONLINE EXCLUSIVE PRICE
    61.00 €
    58.51 € / Pack of 50
    Save 2.49 €
    4% Off
Only null left
Estimated Shipment: 07-11-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Made from food grade, FDA and EC compliant polypropylene
  • Available sterile and non-sterile
  • Wide range of sizes
  • 100% manufactured and packed in a cleanroom
Specifications

Specifications

50 mL
Non-sterile
Single-use
FDA
Low Form
Polypropylene
Autoclavable
Yes
Molded Scale
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.