Learn More
Sampling Systems™ Graduated SteriWare Beakers
This range of graduated beakers are crucial in the production of drugs, chemicals and cosmetics. Individually bagged to prevent cross contamination, these beakers also have a pouring spout to ensure the convenient handling of liquids.
Brand:  Sampling Systems™ 8008P-25S
 View more versions of this product
Product Code. 17982052
  • 114.00 € / Pack of 50
Only null left
Estimated Shipment: 07-11-2025
Add to basket
Description
Description
  • Made from food grade, FDA and EC compliant polypropylene
  • Available sterile and non-sterile
  • Wide range of sizes
  • 100% manufactured and packed in a cleanroom
Specifications
Specifications
25 mL
Sterile
Single-use
FDA
Low Form
Polypropylene
Autoclavable
Yes
Molded Scale