missing translation for 'onlineSavingsMsg'
Learn More

O&M Halyard Sequential Sterilization Wrap (H300)

Halyard Sequential wraps are the industry standard for sterile processing teams.

125.51 € - 250.00 €

Specifications

Product Type Sterilization Wrap
Autoclavable Autoclavable
Certifications/Compliance Meets NFPA Class 1
Color Blue
For Use With (Equipment) Basin sets, instrument sets, procedure trays and large linen packs (6 to 9 lb.)
View More Specs

Products 7
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
30274726
View Documents
O&M
10824
60 x 60 cm (24 x 24 in.)
186.53 €
Pack of 500
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
30274692
View Documents
O&M
10830
76 x 76 cm (30 x 30 in.)
168.12 €
Pack of 300
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
30274731
View Documents
O&M
10836
91 x 91 cm (36 x 36 in.)
239.45 €
Pack of 300
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
30274752
View Documents
O&M
10840
101 x 101 cm (40 x 40 in.)
250.00 €
Pack of 250
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
30274728
View Documents
O&M
10845
114 x 114 cm (45 x 45 in.)
125.51 €
Pack of 100
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
30274765
View Documents
O&M
10848
121 x 121 cm (48 x 48 in.)
146.69 €
Pack of 100
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
30274664
View Documents
O&M
10854
137 x 137 cm (54 x 54 in.)
189.22 €
Pack of 100
Estimated Shipment: 16-05-2025
to see stock.
Add to basket
 
Description

Description

  • Tear-, flame-, lint- and abrasion-resistant
  • POWERGUARD technology for enhanced microbial barrier protection
  • Pre-vacuum steam sterilant penetration
  • Ethylene Oxide (EO) sterilant penetration and residuals
  • Maintenance of package integrity (MPI) to meet FDA requirements
  • Instructions for Use that meet FDA guidelines
  • Class 1 flammability rating
  • Low linting
  • Made in the USA
Specifications

Specifications

Sterilization Wrap
Meets NFPA Class 1
Basin sets, instrument sets, procedure trays and large linen packs (6 to 9 lb.)
Non-sterile
Autoclavable
Blue
SMS Fabric (H300)
Yes
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.