Learn More
Memmert™ HEPA14-Filter For Chamber
HEPA Filter sterile, individually packed, with fixing unit
Brand: Memmert™ ICO105B49800
189.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Includes fixing unit
- According to EN 1822
- Packed in sterile condition
Specifications
Individually packed | |
ICO105 | |
Fixing unit |