missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone-H3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17168-1-AP-150UL
This item is not returnable.
View return policy
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a replication-dependent histone that is a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element.Specifications
| Histone-H3 | |
| Polyclonal | |
| Unconjugated | |
| HIST2H3A | |
| H3/n, H3/o, H3F2, H3FM, HIST2H3A, HIST2H3C, HIST2H3D, histone cluster 2, H3a, Histone H3, Histone H3.2, Histone H3/m, Histone H3/o | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 333932 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q71DI3 | |
| HIST2H3A | |
| Histone-H3 Fusion Protein Ag10644 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction