JT Baker HYDRA-POINT Solvent G BAKER ANALYZED Reagent 2.5L .


Overview
HYDRA-POINT Solvent G BAKER ANALYZED Reagent 2.5L .

Code: NEW

Additional Details:
Additional Details: Weight: 2.50000kg Transport: UN number : 3286 Chem class : 3 Pack group : II


Product Code. 10189160

Quantity Price
1 213.0  € / 2.50 L
Estimated Shipment
22-12-2025
Add to basket


  • Alternative Products