Memmert™ IQ/OQ Document with Device-Specific Works Test Data - Temperature and Humidity
For one free-selectable temperature and humidity value, Includes temperature distribution survey at Memmert to DIN 12880:2007-05
Additional Details:
Additional Details: Weight: 0.01100kg
Product Code. 17159774
Quantity Price
1 3385.0  € / Each
Estimated Shipment
01-08-2025
Add to basket
Description and Specification
Specification
For Use With (Equipment) HPP260eco
Product Type IQ/OQ Document with Device-specific Works Test Data
  • 27 measuring points (sizes 110 - 1060, 2200)
  • 26 measuring points (size 1400)
  • PQ checklist as support for validation by customer