missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCT1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20139-1-AP-150UL
This item is not returnable.
View return policy
Description
SLC16A1 is a monocarboxylate transporter (MCT1) that mediates the movement of lactate and pyruvate across cell membranes. Import and export of these substrates by tissues such as erythrocytes, muscle, intestine, and kidney are ascribed largely to the action of a proton-coupled MCT.Specifications
| MCT1 | |
| Polyclonal | |
| Unconjugated | |
| SLC16A1 | |
| MCT, MCT 1, MCT1, Monocarboxylate transporter 1, SLC16A1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6566 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P53985 | |
| SLC16A1 | |
| MCT1 Fusion Protein Ag14098 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction