| Product Code | Brand | Quantity | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
|---|---|---|---|---|---|---|---|---|
|
30130303
|
Waternation
MLG1-100/10
|
10 x 100 mL |
113.4 €
1 L |
|
||||
|
30130297
|
Waternation
MLG1-500/10
|
10 x 500 mL |
243.6 €
5 L |
|
||||
|
30130300
|
Waternation
MLG1-1000/6
|
6 x 1000 mL |
291.9 €
6 L |
|
Molecular Biological Water, Waternation
Molecular Biological Water is nuclease free, endotoxin and particle free water. Complies with the requirements of ASTM D1193-06.
113.4 € - 291.9 €
Chemical Identifiers
| CAS | 7732-18-5 |
|---|---|
| Molecular Formula | H2O |
| Molecular Weight (g/mol) | 18.02 |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| IUPAC Name | water |
| SMILES | O |
Specification
| pH | 7 |
|---|---|
| CAS | 7732-18-5 |
| Conductivity | 0.1 μS/cm |
| Molecular Formula | H2O |
| InChI Key | XLYOFNOQVPJJNP-UHFFFAOYSA-N |
| SMILES | O |
| IUPAC Name | water |
| Molecular Weight (g/mol) | 18.02 |
| Grade | Molecular Biology |
| Packaging | PET Plastic Bottle |
| Certificate of Analysis | Yes |
| Quantity | 10 x 100 mL |
| For Use With (Application) | Antibody Testing |