missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Human Albumin, 30%
Shop All MP Biomedicals Products
Click to view available options
:
Human Albumin; 50mL
Description
- CAS Number: 70024-90-7
- EC Number: 274-272-6
- MDL Number: MFCD00081418
- Format: solution
- Isoelectric point: 4.7
- Supplied at pH 7, without any preservative
- Sterile with purity >96%
For immunoassays and mammalian cell culture
Specifications
Specifications
| Concentration | 30g/dL |
| For Use With (Application) | Immunoassay, Mammalian Cell Culture |
| Formulation | Supplied at a pH 7.1 ±0.3 and contains 0.1% Sodium Azide as a preservative |
| Name | Human Albumin (30% Solution) |
| pH Range | 7.1 |
| Quantity | 50 mL |
| Source | Human |
| Storage Requirements | 2° to 8°C |
| Form | Solution |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction