missing translation for 'onlineSavingsMsg'
Learn More

Mutlivapor P-12, 230 V, Buchi standard, S-condenser, 1 L

Brand:  Buchi MP11001S10

Mutlivapor P-12, 230 V, Buchi standard, S-condenser, 1 L

Code : 61

Additional Details : Weight : 36.70000kg

Product Code. 10449884

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers