Macherey-Nagel™ N9 Polypropylene Black Screw Caps
Additional Details:
Weight:
0.50000kg
Specification
| Color | Black |
|---|---|
| Material | Polypropylene,PTFE,Silicone |
| Material | Polypropylene,PTFE,Silicone |
| Material | Polypropylene,PTFE,Silicone |
| Septum | Silicone white/PTFE blue, slit |
| Thickness (Metric) | 1 mm |
| Closure Size | N9 |