Learn More
Thermo Scientific™ 96 Well White Plate, Non-Treated Surface, No Lid, Non-Sterile, Pack of 10

Ensure consistent and reliable results using black and white plates optimized for robotics, plate readers and liquid handling systems.
Brand: Thermo Scientific™ 236108
380.92 EUR valid until 2025-08-31
Use promo code "24124" to get your promotional price.
Description
Ensure consistent and reliable results using the high quality Thermo Scientific™ Nunc™ F96 MicroWell™ Black and White Polystyrene Plate. Black and white plates are optimized for robotics, plate readers and liquid handling systems and designed for key imaging characteristics. Available with or without cell culture treatment.
- Background, autoflorescence and crosstalk are improved when you use Nunc Black or White microplates
- Black microplates are recommended for florescence measurements; with minimum back-scattered light and background fluorescence
- White microplates are recommended for luminescence measurements; with maximum reflection and minimal autoluminescence
- Working volume range: 50 to 250μL/well
- Choice of Thermo Scientific™ Nunclon™ Delta surface for cell-based assays
- Customized bar coding is available

Specifications
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) | |
Without Lid | |
10 Pk. | |
White | |
Low Flange Pinchbar | |
96 | |
Non-sterile | |
Flat Bottom (F-well) | |
10 |
Standard | |
8 x 12 | |
Nunc | |
Microplate | |
Polystyrene | |
50 to 250 μL | |
Non-treated | |
180 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.