Learn More
Thermo Scientific™ Nunc™ MicroWell™ 96-Well, Nunclon Delta-Treated, Flat-Bottom Microplate
BIOPHARMA
Color: Clear; Sterility: Sterile; Lid: Without Lid
174.00€ - 418.00€
Specifications
Barcode Custom Barcoding Available
Certifications/Compliance All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
Lid Without Lid
Array 8 x 12
Product Line Nunc
View More Specs
Products 2
Product Code Brand No. of Wells No. per Pack No. per Case Price Quantity & Availability  
Product Code Brand No. of Wells No. per Pack No. per Case Price Quantity & Availability  
10058820
BIOPHARMA
View accessory products View Documents Promotion Details
Thermo Scientific™
168055
96 1 Microplate 50 Microplates
174.00€
Pack of 50
View alternative products
Save Up to 
»
Only null left
In Stock
Edge
Add to basket
10214171
BIOPHARMA
View accessory products View Documents
Thermo Scientific™
156545
96 10 Microplates 180 Microplates
418.00€
Pack of 180
View alternative products
Save Up to 
»
Only null left
In Stock
Add to basket
Description
Description
Thermo Scientific™ Nunc MicroWell 96-Well Microplates are optimized for robotics, automated readers and liquid handling systems. 96-well microplates are used for antibiotic screens, cell-based assays and screening compounds. The flat bottom shape is ideal for microscopic and optical measurements. Our plates are high quality for consistent and reliable results.
  • Clear polystyrene has excellent optical properties making it ideal for optical measurements
  • Untreated polystyrene is hydrophobic and is suited for growth of suspension cultures that can proliferate and grow without attachment
  • Untreated surfaces are ideal for adding coatings in your lab
  • Cell Culture Treated: The Nunclon™ Delta cell culture treated surface is a hydrophilic surface that promotes cell attachment and growth
  • Plates are optimized for robotics, plate readers and liquid handlers
  • Customized bar coding is available
Specifications
Specifications
Custom Barcoding Available
Without Lid
Nunc
Microplate
Polystyrene
96
Nunclon Delta
All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004)
8 x 12
Clear
High Flange
400 μL
Sterile
Flat Bottom (F-well)