| Product Code | Brand | Quantity | Price | ??product.searchResults.label.quantityAvail?? |
|
|||
|---|---|---|---|---|---|---|---|---|
|
16885275
|
Proteintech
26951-1-AP-20UL
|
20 μL |
145.0 €
20µL |
|
||||
|
16875275
|
Proteintech
26951-1-AP-150UL
|
150 μL |
410.0 €
150µL |
|
Perilipin 5 Rabbit anti-Human, Mouse, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.0 € - 410.0 €
Specification
| Antigen | Perilipin 5 |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Immunohistochemistry (Paraffin),Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Specification
| Antigen | Perilipin 5 |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Immunohistochemistry (Paraffin),Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Target Species | Human,Mouse,Pig |
| Formulation | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Gene Accession No. | Q00G26, Q8BVZ1 |
| Gene Alias | LSDP5, OXPAT, Perilipin 5, PLIN5 |
| Gene ID (Entrez) | 100142670, 440503, 66968 |
| Gene Symbols | PLIN5 |
| Immunogen | Perilipin 5 Fusion Protein Ag25272 |
| Isotype | IgG |
| Primary or Secondary | Primary |
| Purification Method | Antigen Affinity Chromatography |
| Content And Storage | -20°C |
| Product Type | Antibody |
| Gene | PLIN5 |