missing translation for 'onlineSavingsMsg'
Learn More
Learn More
pH 11.00 Buffer Solution at 25°C, Reagecon
Shop All Reagecon Products

Click to view available options
Quantity:
500 mL
Unit Size:
500mL
Description
- pH Buffer Reference Standard, pH Buffer Reference Material
- Tested and certified by an ISO17025 Accredited Test Method (INAB Ref: 264T) and NIST traceable.
- Guaranteed stability throughout its entire shelf life, even after opening the bottle.
Specifications
Specifications
| Description | Accuracy ± 0.05 pH units |
| pH | 11.00 ±0.05 |
| Physical Form | Liquid |
| Quantity | 500 mL |
| Packaging | HDPE Bottle GL45 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction