Code:
41
Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Wash Bottle
Manufactured from polypropylene. Bel Art™ Polyethylene Wide Mouth Wash Bottle feature a wide mouth 53mm (2 in.) for easy refilling.
Additional Details:
Weight:
0.01000kg
Specification
Capacity (Metric) | 500 mL |
---|---|
Color | Translucent |
Diameter (Metric) Outer Neck | 53mm |
Includes | Green Polypropylene Cap |
Material | Polyethylene |
Quantity | 4Pack |
Closure Color | Green |
Certifications/Compliance | Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200 |
Mouth | Wide |
Product Type | Wash Bottle |
Chemical Label | Ethyl Acetate |
Closure Material | Polypropylene |
Closure Type | Green Polypropylene Cap |