missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ Polyethylene Wide Mouth Wash Bottle

Manufactured from polypropylene. Bel Art™ Polyethylene Wide Mouth Wash Bottle feature a wide mouth 53mm (2 in.) for easy refilling.

Brand:  Bel-Art™ F11716-0010

 View more versions of this product

Product Code. 12348118

  • 50.84 € / Pack of 4
Only null left
Estimated Shipment: 08-10-2025
Add to basket
This item is not returnable. View return policy

Description

Description

  • Pre-printed with the Chemical name and formula, the National Fire Protection Association (NFPA) Section 704 Four-color Diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment
  • Translucent LDPE bottles; Sodium Hypochlorite is white LDPE for light-protection between 230 and 450nm; Toluene is red LDPE bottle
  • Color-coded polypropylene closures aid in quick and easy identification
  • Provides compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
  • 500ml - 4/bag, 1000ml - 4/bag; assortment is 6 per bag, includes one each: Acetone, Bleach, Distilled Water, Ethanol, Isopropanol, and LYOB

Note: Use LYOB only with solvents compatible with LDPE

Specifications

Specifications

500 mL
53mm
Polyethylene
Natural
Wide
Machine Oil
Natural Polypropylene Cap
Translucent
Natural Polypropylene Cap
4Pack
Compliance with OSHA requirements outlined in Hazard Communication Standard 29 CFR 1910.1200
Wash Bottle
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.