missing translation for 'onlineSavingsMsg'
Learn More

Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Product Code. 15603010 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
5 kg
6 x 100 g
6 x 250 g
6 x 500 g
4 x 5 kg
50 kg
6 x 1 kg
Unit Size:
100g
1kg
250g
500g
50kg
5kg
Pack of 4
Pack of 6
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specifications

Boiling Point 1,323°C
Melting Point 681°C (literature)
CAS Max % 100.0000%
Quantity 6 x 1 kg
Linear Formula KI
Formula Weight 166
Percent Purity >99.5%
Grade puriss. p.a.
Packaging Poly Bottle
Density 3.12 g/cm3
Chemical Name or Material Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.