Honeywell Fluka Potassium iodide, Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%, Honeywell Fluka™

Puriss. p.a., Reag. ISO, Reag. Ph. Eur.,≥99.5%

Overview

Code: NEW

Additional Details:
Additional Details: CAS Number: 7681-11-0 Weight: 50.00000kg


Quantity 50 kg

Product Code. 15673000

Quantity Price
1 7878.85  € / 50kg
Estimated Shipment
16-09-2025
Add to basket


  • Description and Specification
  • Alternative Products
Description and Specification

Chemical Identifiers

CAS 7681-11-0
Molecular Formula IK
Molecular Weight (g/mol) 166.003
MDL Number MFCD00011405
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
PubChem CID 4875
ChEBI CHEBI:8346
IUPAC Name potassium;iodide
SMILES [K+].[I-]

Specification

Boiling Point 1,323°C
Chemical Name or Material Potassium iodide
Melting Point 681°C (literature)
Name Note Puriss. p.a., Reag. ISO, Reag. Ph. Eur., ≥99.5%
CAS 7681-11-0
CAS Max % 100.0000%
Quantity 50 kg
Molecular Formula IK
Linear Formula KI
MDL Number MFCD00011405
UN Number NONH for all modes of transport
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
Synonym potassium iodide,potassium iodide ki,pima,kali iodide,thyro-block,knollide,kisol,asmofug e,potassium diiodide,potassium monoiodide
InChI Key NLKNQRATVPKPDG-UHFFFAOYSA-M
SMILES [K+].[I-]
IUPAC Name potassium;iodide
Molecular Weight (g/mol) 166.003
PubChem CID 4875
ChEBI CHEBI:8346
Formula Weight 166
Percent Purity >99.5%
Grade puriss. p.a.
Packaging Fiberboard Box
Vapor Pressure 1mmHg (745°C)
Density 3.12 g/cm3

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Alternative Products